siRNA Details
Gene Symbol: COA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012985.2 | NP_001013003.1 | 3 | 380 | ||
NM_001206641.2 | NP_001193570.2 | 3 | 464 | |||
NM_001301733.1 | NP_001288662.1 | 2 | 751 | |||
GenBank mRNA | BC025793.1 | - | 3 | 287 | ||
BC035925.1 | - | 3 | 377 | |||
BC116455.1 | - | 3 | 380 | 4540930 | ||
BG195363.1 | - | 1 | 67 | |||
BG204581.1 | - | 2 | 223 | |||
CB136291.1 | - | 3 | 433 | |||
HY169996.1 | - | 3 | 467 |
