siRNA Details
Gene Symbol: C9orf152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012993.2 | NP_001013011.2 | 2 | 1419 | ||
GenBank mRNA | AK290278.1 | - | 2 | 828 | ||
BC110807.1 | - | 2 | 810 | 5178248 | ||
BX648492.1 | - | 2 | 820 | |||
BX648620.1 | - | 2 | 839 |
