siRNA Details
Gene Symbol: Ddx50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013198.2 | NP_001013216.2 | 13, 14 | 1992 | ||
GenBank mRNA | BC091427.1 | - | 7, 8 | 878 | 7366491 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_053183, BC090996, AK030239, AK041281, AF334104.
Gene Ontology Categories:
