siRNA Details
Gene Symbol: Itgb3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013213.1 | NP_001013231.1 | 3 | 440 | ||
XM_008763883.1 | XP_008762105.1 | 4 | 182 | |||
XM_008763884.1 | XP_008762106.1 | 3 | 136 | |||
GenBank mRNA | BC086363.1 | - | 3 | 440 | 7304501 |
