siRNA Details
Gene Symbol: Triobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013220.2 | NP_001013238.2 | 12 | 4408 | ||
XM_006242010.2 | XP_006242072.1 | 3 | 247 | |||
XM_008765758.1 | XP_008763980.1 | 3 | 511 | |||
XM_008765759.1 | XP_008763981.1 | 3 | 511 | |||
XM_008765760.1 | XP_008763982.1 | 3 | 247 | |||
GenBank mRNA | BC088419.1 | - | 3 | 223 | ||
BC169041.1 | - | 3 | 187 | |||
EF203432.1 | - | 2 | 97 | |||
EF203433.1 | - | 2 | 97 |
