siRNA Details
Gene Symbol: Pcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013223.1 | NP_001013241.1 | 6 | 551 | ||
XM_006242417.2 | XP_006242479.1 | 6 | 561 | |||
XM_006242418.2 | XP_006242480.1 | 6 | 561 | |||
XM_006242419.2 | XP_006242481.1 | 6 | 483 | |||
XM_006242420.2 | XP_006242482.1 | 6 | 561 | |||
XM_006242421.2 | XP_006242483.1 | 6 | 561 | |||
XM_006242422.2 | XP_006242484.1 | 6 | 561 | |||
XM_006242423.2 | XP_006242485.1 | 6 | 561 | |||
XM_006242424.2 | XP_006242486.1 | 6 | 981 | |||
XM_006242425.2 | XP_006242487.1 | 6 | 561 | |||
XM_006242426.2 | XP_006242488.1 | 6 | 561 | |||
XM_006242427.2 | XP_006242489.1 | 6 | 561 | |||
XM_008765770.1 | XP_008763992.1 | 6 | 483 | |||
XM_008765771.1 | XP_008763993.1 | 6 | 483 | |||
GenBank mRNA | BC078906.1 | - | 6 | 551 | 7112287 | |
BC078909.1 | - | 5 | 390 | 7127139 | ||
BC097410.1 | - | 7 | 666 | 7454176 | ||
FQ216778.1 | - | 6 | 452 | |||
FQ220695.1 | - | 2 | 129 | |||
FQ231388.1 | - | 2 | 104 |

More Information
Other Species Targeted

Gene Ontology Categories:
