siRNA Details
Gene Symbol: Pcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013223.1 | NP_001013241.1 | 9, 10 | 913 | ||
XM_006242417.2 | XP_006242479.1 | 9, 10 | 923 | |||
XM_006242418.2 | XP_006242480.1 | 9, 10 | 911 | |||
XM_006242419.2 | XP_006242481.1 | 9, 10 | 833 | |||
XM_006242420.2 | XP_006242482.1 | 9, 10 | 911 | |||
XM_006242421.2 | XP_006242483.1 | 9, 10 | 923 | |||
XM_006242422.2 | XP_006242484.1 | 9, 10 | 911 | |||
XM_008765770.1 | XP_008763992.1 | 9, 10 | 845 | |||
GenBank mRNA | BC078906.1 | - | 9, 10 | 913 | 7112287 | |
BC078909.1 | - | 8, 9 | 740 | 7127139 | ||
FQ231388.1 | - | 5, 6 | 454 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011042, NM_001103165, AK169268, EU604548, AK012297, and others.
Gene Ontology Categories:
