siRNA Details
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013227.2 | NP_001013245.1 | 2, 3 | 199 | ||
GenBank mRNA | BC063182.1 | - | 2, 3 | 203 | 6887869 | |
FQ215552.1 | - | 2, 3 | 198 |

More Information
Other Species Targeted

Gene Ontology Categories:
