siRNA Details
Gene Symbol: Nkain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013411.2 | NP_001013429.2 | 7 | 1046 | ||
GenBank mRNA | AY940478.1 | - | 7 | 842 | ||
DQ065607.1 | - | 7 | 979 | |||
EF058049.2 | - | 7 | 979 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
