siRNA Details
Gene Symbol: Arl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013433.1 | NP_001013451.1 | 2 | 830 | ||
XM_006252135.2 | XP_006252197.1 | 3 | 1597 | |||
XM_008770766.1 | XP_008768988.1 | 3 | 1592 | |||
GenBank mRNA | BC091183.1 | - | 2 | 830 | 7365828 |
