siRNA Details
Gene Symbol: YTHDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017798.3 | NP_060268.2 | 4 | 1748 | ||
GenBank mRNA | AB055518.1 | - | 1 | 1089 | ||
AK000398.1 | - | 2 | 909 | |||
AK095795.1 | - | 4 | 1466 | |||
AK300073.1 | - | 2 | 1478 | |||
AL834366.1 | - | 1 | 869 | |||
BC003681.1 | - | 3 | 1422 | |||
BC016920.2 | - | 1 | 1165 | |||
BC025264.1 | - | 1 | 1285 | |||
BC050284.1 | - | 4 | 1748 | 5298438 |
