siRNA Details
Gene Symbol: Kpna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013774.2 | NP_001013796.2 | 4 | 341 | ||
GenBank mRNA | AY950703.1 | - | 4 | 342 | ||
FJ717332.1 | - | 2 | 108 | |||
FJ717333.1 | - | 2 | 108 |
