siRNA Details
Gene Symbol: Fbxw24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013776.4 | NP_001013798.2 | 10 | 1377 | ||
GenBank mRNA | BC080701.1 | - | 10 | 1378 | 30473126 | |
CO808277.1 | - | 5 | 724 | |||
GQ418172.1 | - | 10 | 1378 |
