siRNA Details
Gene Symbol: Fam169b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013811.2 | NP_001013833.1 | 4 | 306 | ||
GenBank mRNA | BC039209.1 | - | 4 | 332 | 4161789 | |
BC039945.1 | - | 4 | 343 | 4161789 |

More Information
Other Species Targeted

Gene Ontology Categories:
