siRNA Details
Gene Symbol: Gm5665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139826.1 | - | 933 | |||
BC032201.1 | - | 545 |
Gene Symbol: BC052688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131969.1 | - | 4 | 633 | ||
AK167253.1 | - | 4 | 629 | |||
BC052688.1 | - | 4 | 588 |
