siRNA Details
Gene Symbol: Paip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014148.1 | NP_001014170.1 | 4 | 391 | ||
XM_008772044.1 | XP_008770266.1 | 2 | 238 | |||
XM_008772046.1 | XP_008770268.1 | 2 | 272 | |||
XM_008772047.1 | XP_008770269.1 | 3 | 294 | |||
XM_008772048.1 | XP_008770270.1 | 3 | 311 | |||
XM_008772049.1 | XP_008770271.1 | 2 | 384 | |||
GenBank mRNA | BC079261.1 | - | 4 | 391 | 7133501 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026420, AK169411, AK009909, AK005462, AK150957, and others.
Gene Ontology Categories:
