siRNA Details
Gene Symbol: Acsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014162.4 | NP_001014184.3 | 14 | 1949 | ||
XM_006230123.1 | XP_006230185.1 | 15 | 1892 | |||
XM_006230126.1 | XP_006230188.1 | 15 | 1892 | |||
GenBank mRNA | BC078915.1 | - | 14 | 1948 | 7096091 |

More Information
Other Species Targeted

Gene Ontology Categories:
