siRNA Details
Gene Symbol: Acsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014162.4 | NP_001014184.3 | 14 | 2344 | ||
XM_006230126.1 | XP_006230188.1 | 15 | 2287 | |||
GenBank mRNA | BC078915.1 | - | 14 | 2343 | 7096091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
