siRNA Details
Gene Symbol: LOC361985 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014174.1 | NP_001014196.1 | 6 | 1226 | ||
GenBank mRNA | BC083693.1 | - | 6 | 1226 | 7189519 | |
BC098673.1 | - | 7 | 1262 | 7130789 | ||
BC105817.1 | - | 7 | 1264 | 7368048 | ||
FQ212139.1 | - | 5 | 1125 | |||
FQ213213.1 | - | 5 | 1161 | |||
FQ214790.1 | - | 7 | 1299 |
