siRNA Details
Gene Symbol: NHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017838.3 | NP_060308.1 | 2, 3 | 371 | ||
GenBank mRNA | AF161404.1 | - | 2, 3 | 281 | ||
AF401219.1 | - | 2, 3 | 229 | |||
AJ293309.1 | - | 2, 3 | 295 | |||
AK000486.1 | - | 2, 3 | 277 | |||
BC000009.2 | - | 2, 3 | 279 | 3505294 | ||
BC006387.2 | - | 2, 3 | 305 | 4109843 | ||
BE797477.1 | - | 2, 3 | 391 | |||
CR457238.1 | - | 2, 3 | 228 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
