siRNA Details
Gene Symbol: Tmem123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014205.1 | NP_001014227.1 | 4 | 1657 | ||
GenBank mRNA | BC089103.1 | - | 4 | 1657 | 7307876 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014205.1 | NP_001014227.1 | 4 | 1657 | ||
GenBank mRNA | BC089103.1 | - | 4 | 1657 | 7307876 |