siRNA Details
Gene Symbol: LOC363337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014221.1 | NP_001014243.1 | 3 | 409 | ||
XM_006255363.2 | XP_006255425.1 | 3 | 465 | |||
XM_008772412.1 | XP_008770634.1 | 3 | 465 | |||
GenBank mRNA | BC082068.1 | - | 3 | 409 | 7132781 |
Gene Symbol: LOC102552540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255351.2 | XP_006255413.1 | 3 | 583 |
Gene Symbol: LOC102557241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772245.1 | XP_008770467.1 | 1 | 80 |
Gene Symbol: LOC679711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758655.1 | XP_008756877.1 | 3 | 498 | ||
XM_008758656.1 | XP_008756878.1 | 3 | 487 | |||
XM_008758657.1 | XP_008756879.1 | 3 | 498 |
Gene Symbol: RGD1562660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255382.2 | XP_006255444.1 | 3 | 569 | ||
XM_006255383.2 | XP_006255445.1 | 3 | 498 | |||
XM_008772436.1 | XP_008770658.1 | 3 | 566 |
