siRNA Details
Gene Symbol: LOC363337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014221.1 | NP_001014243.1 | 3 | 588 | ||
GenBank mRNA | BC082068.1 | - | 3 | 588 | 7132781 |
Gene Symbol: LOC679711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758657.1 | XP_008756879.1 | 3 | 677 |
