siRNA Details
Gene Symbol: LOC290071 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC088274.1 | - | 6, 7 | 854 | ||
BC088276.1 | - | 6, 7 | 836 | 7381661 | ||
BC091200.1 | - | 6, 7 | 888 | |||
BC091267.1 | - | 7, 8 | 821 | 7380098 | ||
BC091273.1 | - | 6, 7 | 998 | 7383056 | ||
BC092593.1 | - | 6, 7 | 868 | 7379916 | ||
BC092595.1 | - | 6, 7 | 971 | 7382388 | ||
BC092596.1 | - | 6, 7 | 836 | 7382760 | ||
BC092597.1 | - | 6, 7 | 825 | |||
BC093389.1 | - | 6, 7 | 822 | 7387331 | ||
BC093392.1 | - | 6, 7 | 873 | 7388480 | ||
M18853.1 | - | 6, 7 | 801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
