siRNA Details
Gene Symbol: LOC290071 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC088274.1 | - | 7 | 1107 | ||
BC088276.1 | - | 7 | 1089 | 7381661 | ||
BC091200.1 | - | 7 | 1141 | |||
BC091267.1 | - | 8 | 1074 | 7380098 | ||
BC091273.1 | - | 7 | 1251 | 7383056 | ||
BC092593.1 | - | 7 | 1121 | 7379916 | ||
BC092595.1 | - | 7 | 1224 | 7382388 | ||
BC092596.1 | - | 7 | 1089 | 7382760 | ||
BC092597.1 | - | 7 | 1078 | |||
BC093389.1 | - | 7 | 1075 | 7387331 | ||
BC093392.1 | - | 7 | 1126 | 7388480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
