siRNA Details
Gene Symbol: Cpsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014245.2 | NP_001014267.1 | 4 | 553 | ||
XM_006231061.2 | XP_006231123.1 | 6 | 708 | |||
XM_006231062.2 | XP_006231124.1 | 5 | 696 | |||
XM_008760238.1 | XP_008758460.1 | 5 | 627 | |||
XM_008760239.1 | XP_008758461.1 | 5 | 627 | |||
XM_008760240.1 | XP_008758462.1 | 4 | 514 | |||
GenBank mRNA | BC083864.1 | - | 4 | 556 | 7127736 |
