siRNA Details
Gene Symbol: RGD1305207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014267.1 | NP_001014289.1 | 2 | 64 | ||
GenBank mRNA | BC079419.1 | - | 2 | 64 | 7113741 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1305207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014267.1 | NP_001014289.1 | 2 | 64 | ||
GenBank mRNA | BC079419.1 | - | 2 | 64 | 7113741 |