siRNA Details
Gene Symbol: Armcx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014274.1 | NP_001014296.1 | 5 | 828 | ||
XM_008773429.1 | XP_008771651.1 | 5 | 938 | |||
GenBank mRNA | BC081886.1 | - | 5 | 828 | 7108660 |
