siRNA Details
Gene Symbol: Kpna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014792.1 | NP_001014792.1 | 17 | 1965 | ||
XM_006252127.2 | XP_006252189.1 | 17 | 2059 | |||
XM_008770761.1 | XP_008768983.1 | 17 | 1914 | |||
GenBank mRNA | AY779027.1 | - | 17 | 1965 |
