siRNA Details
Gene Symbol: Kpna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014793.1 | NP_001014793.1 | 4, 5 | 233 | ||
GenBank mRNA | AY779028.1 | - | 4, 5 | 233 | ||
FQ224515.1 | - | 234 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008467, AK147920, AF020771, BC052162, BC026821, and others.
Gene Ontology Categories:
