siRNA Details
Gene Symbol: Stk38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015025.1 | NP_001015025.1 | 4 | 590 | ||
XM_006256139.2 | XP_006256201.1 | 13 | 1530 | |||
XM_006256140.2 | XP_006256202.1 | 12 | 1449 | |||
XM_006256141.2 | XP_006256203.1 | 12 | 1496 | |||
GenBank mRNA | BC091415.1 | - | 4 | 590 | 7365181 | |
FQ216844.1 | - | 12 | 1379 | |||
FQ217790.1 | - | 12 | 1379 |
