siRNA Details
Gene Symbol: LOC361346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017462.1 | NP_001017462.1 | 4 | 832 | ||
XM_006254929.2 | XP_006254991.1 | 2 | 200 | |||
XM_008772154.1 | XP_008770376.1 | 2 | 443 | |||
GenBank mRNA | BC082051.1 | - | 4 | 832 | 7132090 |
