siRNA Details
Gene Symbol: LOC361635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017463.1 | NP_001017463.1 | 19, 20 | 1655 | ||
XM_006230122.2 | XP_006230184.1 | 19, 20 | 1772 | |||
GenBank mRNA | BC083805.1 | - | 19, 20 | 1655 | 7109216 |
