siRNA Details
Gene Symbol: Btbd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017464.2 | NP_001017464.1 | 3, 4 | 405 | ||
XM_006230403.2 | XP_006230465.1 | 3, 4 | 505 | |||
XM_008759903.1 | XP_008758125.1 | 3, 4 | 505 | |||
XM_008759905.1 | XP_008758127.1 | 3, 4 | 505 | |||
XM_008759906.1 | XP_008758128.1 | 3, 4 | 505 | |||
GenBank mRNA | BC079317.1 | - | 3, 4 | 408 | 7135047 |
