siRNA Details
Gene Symbol: Cep70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017470.1 | NP_001017470.1 | 6, 7 | 646 | ||
XM_006243635.1 | XP_006243697.1 | 6, 7 | 712 | |||
XM_006243639.1 | XP_006243701.1 | 6, 7 | 712 | |||
XM_006243640.2 | XP_006243702.1 | 4, 5 | 487 | |||
XM_006243641.1 | XP_006243703.1 | 5, 6 | 621 | |||
XM_008766552.1 | XP_008764774.1 | 7, 8 | 754 | |||
XM_008766553.1 | XP_008764775.1 | 7, 8 | 739 | |||
XM_008766554.1 | XP_008764776.1 | 8, 9 | 871 | |||
XM_008766555.1 | XP_008764777.1 | 7, 8 | 754 | |||
XM_008766556.1 | XP_008764778.1 | 1, 2 | 274 | |||
GenBank mRNA | BC087075.1 | - | 6, 7 | 646 | 7134655 |
