siRNA Details
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 88 | ||
AF336879.1 | - | 1 | 112 | |||
AF336882.1 | - | 1 | 148 | |||
AF336883.1 | - | 1 | 97 | |||
AF336884.1 | - | 1 | 157 | |||
AF336886.1 | - | 1 | 85 | |||
AF336887.1 | - | 1 | 142 | |||
AF338190.1 | - | 1 | 130 | |||
AF338193.1 | - | 1 | 148 | |||
AF338194.1 | - | 1 | 151 | |||
AF338195.1 | - | 1 | 148 | |||
AF338196.1 | - | 1 | 160 | |||
AF338198.1 | - | 1 | 148 | |||
AF338199.1 | - | 1 | 115 | |||
AF469044.1 | - | 1 | 97 |
Gene Symbol: FKSG48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336875.1 | - | 1 | 163 |
Gene Symbol: HELLPAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JX088243.1 | - | 1 | 53296 |
Gene Symbol: LOC100506290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336880.1 | - | 1 | 169 |
Gene Symbol: LOC402469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336877.1 | - | 1 | 160 | ||
AF336881.1 | - | 1 | 160 | |||
AF338191.1 | - | 1 | 145 |
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127395.2 | NP_001120867.1 | 4 | 2210 | ||
NM_145280.5 | NP_660323.3 | 4 | 2071 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126330.1 | - | 9 | 2453 |
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024077.1 | - | 4 | 1420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
