siRNA Details
Gene Symbol: SBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024401.2 | NP_001019572.1 | 4 | 4948 | ||
XM_005255315.3 | XP_005255372.1 | 4 | 4982 | |||
GenBank mRNA | AL390134.1 | - | 1 | 2087 | ||
AY874862.2 | - | 4 | 4948 |
