siRNA Details
Gene Symbol: ZNF772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024596.2 | NP_001019767.1 | 5 | 1908 | ||
NM_001144068.1 | NP_001137540.1 | 4 | 1785 | |||
XM_005258943.4 | XP_005259000.1 | 3 | 5038 | |||
XM_005258944.3 | XP_005259001.1 | 2 | 4999 | |||
XM_011526980.2 | XP_011525282.1 | 5 | 5542 | |||
XM_017026813.1 | XP_016882302.1 | 4 | 5419 | |||
GenBank mRNA | AK301562.1 | - | 2 | 1587 | ||
AK302574.1 | - | 4 | 1747 | |||
BX647068.1 | - | 5 | 1908 | |||
CR749225.1 | - | 2 | 1594 |
