siRNA Details
Gene Symbol: Gm5039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094938.1 | - | 1 | 1203 |
Gene Symbol: BB287469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177573.1 | NP_001171044.1 | 2 | 1418 | ||
GenBank mRNA | AK139752.1 | - | 2 | 1418 |
Gene Symbol: Gm2022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177574.1 | NP_001171045.1 | 2 | 1392 | ||
GenBank mRNA | AK146348.1 | - | 1289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
