siRNA Details
Gene Symbol: Gm5039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094938.1 | - | 1 | 1252 |
Gene Symbol: Gm2016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122662.1 | NP_001116134.1 | 2 | 1459 | ||
GenBank mRNA | AK163212.1 | - | 2 | 1459 |
Gene Symbol: Gm5662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013824.3 | NP_001013846.1 | 2 | 1456 | ||
GenBank mRNA | AK146341.1 | - | 2 | 1459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
