siRNA Details
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024871.1 | NP_001020042.1 | 5 | 761 | ||
XM_006233955.1 | XP_006234017.1 | 4 | 808 | |||
XM_006233957.2 | XP_006234019.1 | 4 | 416 | |||
XM_006233958.1 | XP_006234020.1 | 4 | 675 | |||
XM_006233959.2 | XP_006234021.1 | 4 | 486 | |||
XM_006233963.1 | XP_006234025.1 | 4 | 724 | |||
XM_006233964.2 | XP_006234026.1 | 4 | 333 | |||
XM_006233966.1 | XP_006234028.1 | 4 | 720 | |||
XM_006233967.1 | XP_006234029.1 | 4 | 587 | |||
XM_008761724.1 | XP_008759946.1 | 6 | 958 | |||
XM_008761725.1 | XP_008759947.1 | 5 | 516 | |||
XM_008761726.1 | XP_008759948.1 | 5 | 489 | |||
XM_008761727.1 | XP_008759949.1 | 4 | 835 | |||
XM_008761728.1 | XP_008759950.1 | 4 | 444 | |||
XM_008761729.1 | XP_008759951.1 | 4 | 702 | |||
XM_008761730.1 | XP_008759952.1 | 5 | 760 | |||
XM_008761731.1 | XP_008759953.1 | 5 | 405 | |||
XM_008761732.1 | XP_008759954.1 | 5 | 405 | |||
XM_008761733.1 | XP_008759955.1 | 3 | 726 | |||
XM_008761736.1 | XP_008759958.1 | 3 | 378 | |||
XM_008761738.1 | XP_008759960.1 | 4 | 591 | |||
XM_008761739.1 | XP_008759961.1 | 3 | 637 | |||
GenBank mRNA | BC097415.1 | - | 5 | 761 | 7454340 |

More Information
Other Species Targeted

