siRNA Details
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025018.1 | NP_001020189.1 | 10 | 1076 | ||
XM_006233120.2 | XP_006233182.1 | 10 | 1020 | |||
XM_006233121.2 | XP_006233183.1 | 9 | 960 | |||
XM_006233122.2 | XP_006233184.1 | 9 | 1061 | |||
XM_006233123.2 | XP_006233185.1 | 9 | 923 | |||
GenBank mRNA | BC091251.1 | - | 10 | 1076 | 7372838 | |
FQ218349.1 | - | 10 | 1088 | |||
FQ228782.1 | - | 10 | 1173 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK008532, AK009940, BC024458, AK172455, AK012044, and others.
Gene Ontology Categories:
