siRNA Details
Gene Symbol: Stap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025026.1 | NP_001020197.1 | 11 | 1391 | ||
GenBank mRNA | BC091228.1 | - | 11 | 1391 | 7321609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
