siRNA Details
Gene Symbol: Fam35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025028.2 | NP_001020199.2 | 4 | 1524 | ||
XM_008771135.1 | XP_008769357.1 | 3 | 1454 | |||
XM_008771136.1 | XP_008769358.1 | 2 | 1258 | |||
XM_008771137.1 | XP_008769359.1 | 2 | 1222 | |||
XM_008771138.1 | XP_008769360.1 | 4 | 1524 | |||
GenBank mRNA | BC089872.1 | - | 4 | 1524 | 7325236 |
