siRNA Details
Gene Symbol: GYPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304382.1 | NP_001291311.1 | 3 | 201 | ||
NM_002100.5 | NP_002091.3 | 2 | 121 | |||
XM_011531903.2 | XP_011530205.1 | 2 | 256 | |||
XM_011531904.2 | XP_011530206.1 | 3 | 2760 | |||
XM_011531905.2 | XP_011530207.1 | 3 | 408 | |||
XM_017008137.1 | XP_016863626.1 | 2 | 2455 | |||
GenBank mRNA | AY509882.1 | - | 2 | 63 | ||
AY950610.1 | - | 2 | 63 | |||
AY950611.1 | - | 2 | 63 | |||
AY950612.1 | - | 2 | 63 | |||
BC069310.1 | - | 2 | 113 | 7262104 | ||
BC121077.1 | - | 2 | 97 | 40119713 | ||
BC121078.1 | - | 2 | 97 | 40119716 | ||
EU338224.1 | - | 3 | 224 | |||
EU338225.1 | - | 2 | 100 | |||
EU338226.1 | - | 3 | 158 | |||
EU338229.1 | - | 3 | 224 | |||
EU338230.1 | - | 3 | 224 | |||
EU338236.1 | - | 3 | 224 | |||
EU338237.1 | - | 3 | 224 | |||
J02982.1 | - | 2 | 73 | |||
X08055.1 | - | 4 | 199 |
Gene Symbol: GYPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308187.1 | NP_001295116.1 | 2 | 214 | ||
NM_002099.7 | NP_002090.4 | 2 | 214 | |||
XM_017008134.1 | XP_016863623.1 | 2 | 218 | |||
XM_017008135.1 | XP_016863624.1 | 3 | 281 | |||
GenBank mRNA | AA334250.1 | - | 2 | 100 | ||
BC005319.1 | - | 2 | 120 | 3930700 | ||
EU338233.1 | - | 3 | 183 | |||
S77082.1 | - | 1 | 36 | |||
U00177.1 | - | 2 | 63 | |||
U00178.1 | - | 2 | 63 | |||
X08054.1 | - | 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
