siRNA Details
Gene Symbol: ZNF708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297560.1 | NP_001284489.1 | 4 | 3791 | ||
NM_001297561.1 | NP_001284490.1 | 3 | 3582 | |||
NM_021269.2 | NP_067092.2 | 4 | 3707 | |||
XM_017027202.1 | XP_016882691.1 | 4 | 3732 | |||
XM_017027203.1 | XP_016882692.1 | 2 | 3512 | |||
XM_017027204.1 | XP_016882693.1 | 5 | 3273 | |||
GenBank mRNA | AK128184.1 | - | 5 | 3533 | ||
AK131527.1 | - | 4 | 3707 | |||
BC039403.1 | - | 1 | 330 | |||
BX103920.1 | - | 1 | 404 |
