siRNA Details
Gene Symbol: IMPAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017813.4 | NP_060283.3 | 2 | 762 | ||
GenBank mRNA | AK000428.1 | - | 2 | 759 | ||
AY032885.1 | - | 2 | 476 | |||
BC017797.1 | - | 2 | 711 | 4306100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
