siRNA Details
Gene Symbol: APITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199294.2 | NP_954988.1 | 5 | 860 | ||
NR_036462.1 | 5 | 1048 | ||||
GenBank mRNA | BC029430.1 | - | 5 | 519 | 4051739 | |
BG213105.1 | - | 2 | 261 | |||
BU902753.1 | - | 4 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
