siRNA Details
Gene Symbol: Gm20871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC099537.1 | - | 203 | 30916067 |
Gene Symbol: 1700040F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006653.1 | - | 100 | |||
AK077064.1 | - | 2 | 93 | |||
FJ541086.1 | - | 142 | ||||
FJ541087.1 | - | 74 | ||||
FJ541088.1 | - | 74 | ||||
FJ541089.1 | - | 75 |
Gene Symbol: D630029K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ541076.1 | - | 206 |
Gene Symbol: Gm20736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037748.1 | NP_001032837.2 | 3 | 228 | ||
GenBank mRNA | BC100464.1 | - | 3 | 228 | 6705131 |
Gene Symbol: Gm20857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU945722.1 | - | 231 |
Gene Symbol: Pmel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ541090.1 | - | 74 |
Gene Symbol: Sly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201530.2 | NP_963288.2 | 3 | 213 | ||
GenBank mRNA | BC049626.1 | - | 3 | 213 | 6705850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
