siRNA Details
Gene Symbol: C11orf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142946.1 | NP_001136418.1 | 4 | 412 | ||
NR_024621.1 | 3 | 327 | ||||
XM_011520034.1 | XP_011518336.1 | 4 | 412 | |||
GenBank mRNA | AB029488.1 | - | 3 | 480 | ||
AK290921.1 | - | 3 | 281 | |||
AK310172.1 | - | 4 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
